Assay Details
Gene Symbol: Hnmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031044.3 | NP_112306.1 | 3 - 4 | 312 | 99 | |
GenBank mRNA | BC087635.1 | - | 3 - 4 | 503 | 99 | |
D10693.1 | - | 3 - 4 | 312 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
