Assay Details
Gene Symbol: Sh3glb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009692.1 | NP_001009692.1 | 9 - 10 | 848 | 99 | |
XM_006233816.2 | XP_006233878.1 | 10 - 11 | 975 | 99 | ||
XM_006233817.2 | XP_006233879.1 | 10 - 11 | 975 | 99 | ||
XM_006233818.2 | XP_006233880.1 | 9 - 10 | 824 | 99 | ||
XM_006233819.2 | XP_006233881.1 | 8 - 9 | 812 | 99 | ||
XM_006233820.2 | XP_006233882.1 | 8 - 9 | 812 | 99 | ||
XM_006233822.2 | XP_006233884.1 | 7 - 8 | 749 | 99 | ||
XM_008761661.1 | XP_008759883.1 | 9 - 10 | 963 | 99 | ||
XM_008761662.1 | XP_008759884.1 | 7 - 8 | 749 | 99 | ||
GenBank mRNA | BC087652.1 | - | 9 - 10 | 848 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
