Assay Details
Gene Symbol: Egfl7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139104.1 | NP_620804.1 | 9 - 10 | 1223 | 125 | |
XM_006233668.2 | XP_006233730.1 | 9 - 10 | 1326 | 125 | ||
XM_006233669.2 | XP_006233731.1 | 8 - 9 | 880 | 125 | ||
GenBank mRNA | AF223678.1 | - | 9 - 10 | 1223 | 125 | |
BC078725.1 | - | 9 - 10 | 1167 | 125 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
