Assay Details
Gene Symbol: Gtf3c5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079941.1 | NP_001073410.1 | 8 - 9 | 1416 | 151 | |
XM_006233835.2 | XP_006233897.1 | 8 - 9 | 1425 | 151 | ||
GenBank mRNA | BC129109.1 | - | 8 - 9 | 1416 | 151 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
