Assay Details
Gene Symbol: Ggh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012960.2 | NP_037092.1 | 3 - 4 | 416 | 142 | |
GenBank mRNA | BC087602.1 | - | 3 - 4 | 416 | 142 | |
U38379.1 | - | 3 - 4 | 266 | 142 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
