Assay Details
Gene Symbol: Grhpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113754.1 | NP_001107226.1 | 4 - 5 | 467 | 73 | |
GenBank mRNA | BC158680.1 | - | 4 - 5 | 451 | 73 | |
FQ210026.1 | - | 4 - 5 | 438 | 73 | ||
FQ210631.1 | - | 4 - 5 | 414 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
