Assay Details
Gene Symbol: Polr1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107938.1 | NP_001101408.1 | 5 - 6 | 474 | 101 | |
XM_008763673.1 | XP_008761895.1 | 5 - 6 | 806 | 101 | ||
GenBank mRNA | BC158699.1 | - | 5 - 6 | 462 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
