Assay Details
Gene Symbol: Mms22l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135780.1 | NP_001129252.1 | 3 - 4 | 739 | 140 | |
XM_006237954.2 | XP_006238016.1 | 3 - 4 | 791 | 140 | ||
GenBank mRNA | BC160888.1 | - | 3 - 4 | 696 | 140 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
