Assay Details
Gene Symbol: Ripk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191865.1 | NP_001178794.1 | 10 - 11 | 1415 | 68 | |
GenBank mRNA | BC107468.1 | - | 8 - 9 | 846 | 68 | |
FQ230093.1 | - | 4 - 5 | 382 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
