Assay Details
Gene Symbol: Ubap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012190.1 | NP_001012190.1 | 1 - 2 | 200 | 142 | |
XM_008763626.1 | XP_008761848.1 | 1 - 2 | 209 | 142 | ||
GenBank mRNA | BC083594.1 | - | 1 - 2 | 200 | 142 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC083594.1, NM_001012190.1, XM_008763626.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
