Assay Details
Gene Symbol: Trmt10b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013090.2 | NP_001013108.1 | 3 - 4 | 451 | 67 | |
XM_006238042.1 | XP_006238104.1 | 4 - 5 | 501 | 67 | ||
XM_006238043.1 | XP_006238105.1 | 4 - 5 | 700 | 67 | ||
XM_006238044.2 | XP_006238106.1 | 4 - 5 | 461 | 67 | ||
GenBank mRNA | BC086603.1 | - | 3 - 4 | 455 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
