Assay Details
Gene Symbol: Trmt10b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013090.2 | NP_001013108.1 | 5 - 6 | 689 | 87 | |
XM_006238042.1 | XP_006238104.1 | 6 - 7 | 739 | 87 | ||
XM_006238043.1 | XP_006238105.1 | 6 - 7 | 938 | 87 | ||
XM_006238044.2 | XP_006238106.1 | 6 - 7 | 699 | 87 | ||
GenBank mRNA | BC086603.1 | - | 5 - 6 | 693 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
