Assay Details
Gene Symbol: Il1rl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133575.1 | NP_598259.1 | 3 - 4 | 390 | 146 | |
GenBank mRNA | U49066.1 | - | 3 - 4 | 390 | 146 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
