Assay Details
Gene Symbol: Bmpr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080407.1 | NP_536332.1 | 11 - 12 | 1958 | 131 | |
XM_006244951.2 | XP_006245013.1 | 12 - 13 | 2444 | 131 | ||
XM_006244952.2 | XP_006245014.1 | 12 - 13 | 2444 | 131 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
