Assay Details
Gene Symbol: Il1r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053953.1 | NP_446405.1 | 1 - 2 | 79 | 78 | |
XM_008766983.1 | XP_008765205.1 | 1 - 2 | 464 | 78 | ||
GenBank mRNA | BC091564.1 | - | 1 - 2 | 53 | 78 | |
FQ228742.1 | - | 1 - 2 | 78 | 78 | ||
FQ229102.1 | - | 1 - 2 | 78 | 78 | ||
Z22812.1 | - | 1 - 2 | 79 | 78 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC091564.1, NM_053953.1, XM_008766983.1, Z22812.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
