Assay Details
Gene Symbol: Mfsd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108215.1 | NP_001101685.1 | 4 - 5 | 461 | 73 | |
XM_006244823.2 | XP_006244885.1 | 4 - 5 | 453 | 73 | ||
XM_006244824.2 | XP_006244886.1 | 4 - 5 | 597 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
