Assay Details
Gene Symbol: Mrps9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100549.1 | NP_001094019.1 | 6 - 7 | 585 | 59 | |
XM_008767019.1 | XP_008765241.1 | 6 - 7 | 596 | 59 | ||
GenBank mRNA | BC088242.1 | - | 5 - 6 | 397 | 59 | |
BC158704.1 | - | 6 - 7 | 553 | 59 | ||
CV106852.1 | - | 6 - 7 | 585 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
