Assay Details
Gene Symbol: Hibch | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013112.1 | NP_001013130.1 | 1 - 2 | 136 | 116 | |
XM_006244895.1 | XP_006244957.1 | 1 - 2 | 222 | 116 | ||
GenBank mRNA | BC083737.1 | - | 1 - 2 | 136 | 116 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC083737.1, NM_001013112.1, XM_006244895.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
