Assay Details
Gene Symbol: Ino80d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191809.1 | NP_001178738.1 | 3 - 4 | 1056 | 67 | |
XM_006245023.2 | XP_006245085.1 | 6 - 7 | 1425 | 67 | ||
XM_006245024.2 | XP_006245086.1 | 5 - 6 | 2758 | 67 | ||
XM_006245025.2 | XP_006245087.1 | 5 - 6 | 1321 | 67 | ||
XM_006245026.2 | XP_006245088.1 | 6 - 7 | 1668 | 67 | ||
XM_006245029.2 | XP_006245091.1 | 6 - 7 | 1535 | 67 | ||
XM_006245031.2 | XP_006245093.1 | 5 - 6 | 1431 | 67 | ||
XM_006245032.2 | XP_006245094.1 | 5 - 6 | 1564 | 67 | ||
XM_006245033.2 | XP_006245095.1 | 5 - 6 | 1340 | 67 | ||
XM_006245034.2 | XP_006245096.1 | 5 - 6 | 1230 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
