Assay Details
Gene Symbol: Rab23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109005.1 | NP_001102475.1 | 6 - 7 | 816 | 93 | |
XM_008766955.1 | XP_008765177.1 | 6 - 7 | 1477 | 93 | ||
XM_008766956.1 | XP_008765178.1 | 6 - 7 | 1482 | 93 | ||
XM_008766957.1 | XP_008765179.1 | 6 - 7 | 802 | 93 | ||
XM_008766958.1 | XP_008765180.1 | 6 - 7 | 1144 | 93 | ||
XM_008766959.1 | XP_008765181.1 | 6 - 7 | 1475 | 93 | ||
XM_008766960.1 | XP_008765182.1 | 6 - 7 | 1470 | 93 | ||
XM_008766961.1 | XP_008765183.1 | 4 - 5 | 2193 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
