Assay Details
Gene Symbol: Actr1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039028.2 | NP_001034117.2 | 7 - 8 | 857 | 147 | |
GenBank mRNA | BC099152.1 | - | 2 - 3 | 451 | 147 | |
BC166803.1 | - | 7 - 8 | 824 | 147 | ||
DY315273.1 | - | 4 - 5 | 470 | 147 | ||
FQ228979.1 | - | 7 - 8 | 825 | 147 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
