Assay Details
Gene Symbol: Clk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106913.2 | NP_001100383.2 | 3 - 4 | 518 | 105 | |
GenBank mRNA | CV127206.1 | - | 3 - 4 | 487 | 105 | |
FQ232068.1 | - | 3 - 4 | 528 | 105 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
