Assay Details
Gene Symbol: Cercam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011962.1 | NP_001011962.1 | 5 - 6 | 486 | 70 | |
XM_006233768.2 | XP_006233830.1 | 2 - 3 | 1154 | 70 | ||
XM_006233769.2 | XP_006233831.1 | 3 - 4 | 546 | 70 | ||
XM_006233770.2 | XP_006233832.1 | 5 - 6 | 1526 | 70 | ||
XM_006233771.2 | XP_006233833.1 | 2 - 3 | 1155 | 70 | ||
GenBank mRNA | BC085782.1 | - | 5 - 6 | 486 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
