Assay Details
Gene Symbol: Cercam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011962.1 | NP_001011962.1 | 8 - 9 | 950 | 133 | |
XM_006233768.2 | XP_006233830.1 | 5 - 6 | 1618 | 133 | ||
XM_006233769.2 | XP_006233831.1 | 6 - 7 | 1010 | 133 | ||
XM_006233770.2 | XP_006233832.1 | 8 - 9 | 1990 | 133 | ||
XM_006233771.2 | XP_006233833.1 | 4 - 5 | 1484 | 133 | ||
GenBank mRNA | BC085782.1 | - | 8 - 9 | 950 | 133 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
