Assay Details
Gene Symbol: Dbh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013158.2 | NP_037290.2 | 10 - 11 | 1578 | 101 | |
GenBank mRNA | BF525224.1 | - | 3 - 4 | 316 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
