Assay Details
Gene Symbol: Dbh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013158.2 | NP_037290.2 | 11 - 12 | 1733 | 129 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
cytokine production
blood vessel remodeling
response to amphetamine
leukocyte mediated immunity
catecholamine metabolic process
memory
locomotory behavior
associative learning
visual learning
response to xenobiotic stimulus
response to iron ion
response to ozone
multicellular organism aging
response to organic cyclic compound
response to estradiol
social behavior
response to isolation stress
response to immobilization stress
regulation of cell population proliferation
homoiothermy
dopamine catabolic process
norepinephrine biosynthetic process
glucose homeostasis
fear response
maternal behavior
response to peptide hormone
response to ethanol
positive regulation of vasoconstriction
octopamine metabolic process
response to copper ion
behavioral response to ethanol
response to pain
leukocyte migration
sensory perception of taste
bone development
cellular response to manganese ion
cellular response to nicotine
regulation of extrinsic apoptotic signaling pathway
dopamine beta-monooxygenase activity
copper ion binding
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, reduced ascorbate as one donor, and incorporation of one atom of oxygen
L-ascorbic acid binding
cytokine production
blood vessel remodeling
response to amphetamine
leukocyte mediated immunity
catecholamine metabolic process
memory
locomotory behavior
associative learning
visual learning
response to xenobiotic stimulus
response to iron ion
response to ozone
multicellular organism aging
response to organic cyclic compound
response to estradiol
social behavior
response to isolation stress
response to immobilization stress
regulation of cell population proliferation
homoiothermy
dopamine catabolic process
norepinephrine biosynthetic process
glucose homeostasis
fear response
maternal behavior
response to peptide hormone
response to ethanol
positive regulation of vasoconstriction
octopamine metabolic process
response to copper ion
behavioral response to ethanol
response to pain
leukocyte migration
sensory perception of taste
bone development
cellular response to manganese ion
cellular response to nicotine
regulation of extrinsic apoptotic signaling pathway
dopamine beta-monooxygenase activity
copper ion binding
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, reduced ascorbate as one donor, and incorporation of one atom of oxygen
L-ascorbic acid binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P083610 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962309 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 685516 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144107 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55584 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 58876 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 417816 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 343881 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260903 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 549911 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 769399 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 501801 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 123356 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367876 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 130974 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 53259 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P083610 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P083610 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P083610 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P381686 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 31725 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10834692 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10834700 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10834702 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10834693 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10834694 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10834703 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10834697 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10834695 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10834701 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10834699 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10834696 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10834698 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10834691 | - | - |
Rat Genome 230 2.0 Array | 1370564_at | - | - |
Rat Genome 230 2.0 Array | 1370564_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1370564_at | - | - |
Affymetrix HT Rat Focus Array | 1370564_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0300001316.rn.1 | TC0300000137.rn.1 | RTA TC |
Rat Expression Set 230 | 1370564_at | - | - |
Rat Expression Set 230 | 1370564_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10834691 | - | - |
Rat Genome U34 Set | L12407_at | - | - |
Rat Genome U34 Set | L12407_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21731357 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1374582 | - | - |
RatRef-12 Expression BeadChip | ILMN_66537 | - | - |
