Assay Details
Gene Symbol: Noxa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100171.1 | NP_001093641.1 | 4 - 5 | 505 | 148 | |
XM_006233600.2 | XP_006233662.1 | 4 - 5 | 792 | 148 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
