Assay Details
Gene Symbol: Ubac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007742.1 | NP_001007743.1 | 2 - 3 | 427 | 99 | |
XM_006233689.1 | XP_006233751.1 | 2 - 3 | 454 | 99 | ||
GenBank mRNA | BC083603.1 | - | 2 - 3 | 427 | 99 | |
FQ210767.1 | - | 2 - 3 | 313 | 99 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
