Assay Details
Gene Symbol: Ntmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025019.1 | NP_001020190.1 | 3 - 4 | 585 | 79 | |
XM_006233919.2 | XP_006233981.1 | 4 - 5 | 1410 | 79 | ||
XM_008761670.1 | XP_008759892.1 | 2 - 3 | 475 | 79 | ||
GenBank mRNA | BC091294.1 | - | 3 - 4 | 585 | 79 | |
FQ216821.1 | - | 3 - 4 | 600 | 79 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
