Assay Details
Gene Symbol: Ndor1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107818.1 | NP_001101288.1 | 12 - 13 | 1633 | 58 | |
XM_006233612.2 | XP_006233674.1 | 11 - 12 | 1925 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
