Assay Details
Gene Symbol: Ndor1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107818.1 | NP_001101288.1 | 5 - 6 | 587 | 75 | |
XM_006233612.2 | XP_006233674.1 | 4 - 5 | 879 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
