Assay Details
Gene Symbol: Ndor1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107818.1 | NP_001101288.1 | 8 - 9 | 1136 | 80 | |
XM_006233612.2 | XP_006233674.1 | 7 - 8 | 1428 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
