Assay Details
Gene Symbol: Ak8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004266.1 | NP_001004266.1 | 11 - 12 | 1130 | 62 | |
XM_006233794.2 | XP_006233856.1 | 12 - 13 | 1166 | 62 | ||
GenBank mRNA | BC079446.1 | - | 11 - 12 | 1130 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
