Assay Details
Gene Symbol: Phyhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013081.1 | NP_001013099.1 | 2 - 3 | 236 | 64 | |
XM_006233777.2 | XP_006233839.1 | 3 - 4 | 620 | 64 | ||
XM_006233778.2 | XP_006233840.1 | 3 - 4 | 620 | 64 | ||
XM_006233779.2 | XP_006233841.1 | 3 - 4 | 1313 | 64 | ||
XM_006233780.2 | XP_006233842.1 | 3 - 4 | 376 | 64 | ||
GenBank mRNA | BC091389.1 | - | 2 - 3 | 236 | 64 | |
FQ212963.1 | - | 2 - 3 | 252 | 64 | ||
FQ219402.1 | - | 2 - 3 | 257 | 64 | ||
FQ223356.1 | - | 2 - 3 | 243 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
