Assay Details
Gene Symbol: Phyhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013081.1 | NP_001013099.1 | 3 - 4 | 309 | 105 | |
XM_006233777.2 | XP_006233839.1 | 4 - 5 | 693 | 105 | ||
XM_006233778.2 | XP_006233840.1 | 4 - 5 | 693 | 105 | ||
XM_006233779.2 | XP_006233841.1 | 4 - 5 | 1386 | 105 | ||
XM_006233780.2 | XP_006233842.1 | 4 - 5 | 449 | 105 | ||
GenBank mRNA | BC091389.1 | - | 3 - 4 | 309 | 105 | |
FQ219402.1 | - | 3 - 4 | 330 | 105 | ||
FQ223356.1 | - | 3 - 4 | 316 | 105 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
