Assay Details
Gene Symbol: Phyhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013081.1 | NP_001013099.1 | 5 - 6 | 410 | 62 | |
XM_006233777.2 | XP_006233839.1 | 6 - 7 | 794 | 62 | ||
XM_006233778.2 | XP_006233840.1 | 6 - 7 | 794 | 62 | ||
XM_006233779.2 | XP_006233841.1 | 6 - 7 | 1487 | 62 | ||
XM_006233780.2 | XP_006233842.1 | 6 - 7 | 550 | 62 | ||
GenBank mRNA | BC091389.1 | - | 5 - 6 | 410 | 62 | |
FQ219402.1 | - | 5 - 6 | 431 | 62 | ||
FQ223356.1 | - | 5 - 6 | 417 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
