Assay Details
Gene Symbol: Phyhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013081.1 | NP_001013099.1 | 6 - 7 | 473 | 101 | |
XM_006233777.2 | XP_006233839.1 | 7 - 8 | 857 | 101 | ||
XM_006233778.2 | XP_006233840.1 | 7 - 8 | 857 | 101 | ||
XM_006233779.2 | XP_006233841.1 | 7 - 8 | 1550 | 101 | ||
XM_006233780.2 | XP_006233842.1 | 7 - 8 | 613 | 101 | ||
GenBank mRNA | BC091389.1 | - | 6 - 7 | 473 | 101 | |
FQ212963.1 | - | 6 - 7 | 489 | 101 | ||
FQ219402.1 | - | 6 - 7 | 494 | 101 | ||
FQ219572.1 | - | 3 - 4 | 476 | 101 | ||
FQ223356.1 | - | 6 - 7 | 480 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
