Assay Details
Gene Symbol: Dolpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006233783.2 | XP_006233845.1 | 5 - 6 | 1691 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
