Assay Details
Gene Symbol: Rngtt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107923.2 | NP_001101393.1 | 11 - 12 | 1384 | 110 | |
XM_006237990.2 | XP_006238052.1 | 11 - 12 | 1398 | 110 | ||
XM_006237991.2 | XP_006238053.1 | 11 - 12 | 1370 | 110 | ||
GenBank mRNA | BC168990.1 | - | 11 - 12 | 1380 | 110 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
