Assay Details
Gene Symbol: Rngtt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107923.2 | NP_001101393.1 | 6 - 7 | 797 | 104 | |
XM_006237990.2 | XP_006238052.1 | 6 - 7 | 811 | 104 | ||
XM_006237991.2 | XP_006238053.1 | 6 - 7 | 783 | 104 | ||
GenBank mRNA | BC168990.1 | - | 6 - 7 | 793 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
