Assay Details
Gene Symbol: Tpm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024345.2 | NP_001019516.1 | 8 - 9 | 944 | 65 | |
XM_008763680.1 | XP_008761902.1 | 8 - 9 | 1284 | 65 | ||
GenBank mRNA | BC090009.1 | - | 8 - 9 | 865 | 65 | |
CF110059.1 | - | 8 - 9 | 830 | 65 | ||
FQ217324.1 | - | 8 - 9 | 881 | 65 | ||
FQ219788.1 | - | 8 - 9 | 841 | 65 | ||
FQ219821.1 | - | 8 - 9 | 881 | 65 | ||
FQ220438.1 | - | 8 - 9 | 841 | 65 | ||
FQ220606.1 | - | 8 - 9 | 881 | 65 | ||
FQ220675.1 | - | 8 - 9 | 881 | 65 | ||
FQ229470.1 | - | 8 - 9 | 881 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
