Assay Details
Gene Symbol: Bag1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106647.3 | NP_001100117.2 | 1 - 2 | 539 | 110 | |
NM_001256084.1 | NP_001243013.1 | 1 - 2 | 539 | 110 | ||
GenBank mRNA | BC159418.1 | - | 1 - 2 | 539 | 110 | |
DY309876.1 | - | 1 - 2 | 536 | 110 | ||
FQ215132.1 | - | 1 - 2 | 501 | 110 | ||
FQ224976.1 | - | 1 - 2 | 536 | 110 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
