Assay Details
Gene Symbol: Bag1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106647.3 | NP_001100117.2 | 4 - 5 | 867 | 81 | |
NM_001256084.1 | NP_001243013.1 | 4 - 5 | 867 | 81 | ||
GenBank mRNA | BC159418.1 | - | 4 - 5 | 867 | 81 | |
BF523560.1 | - | 3 - 4 | 277 | 81 | ||
BI280304.1 | - | 3 - 4 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
