Assay Details
Gene Symbol: Bag1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106647.3 | NP_001100117.2 | 6 - 7 | 1036 | 72 | |
NM_001256084.1 | NP_001243013.1 | 6 - 7 | 1036 | 72 | ||
GenBank mRNA | BC159418.1 | - | 6 - 7 | 1036 | 72 | |
BF523560.1 | - | 5 - 6 | 446 | 72 | ||
BI280304.1 | - | 1 - 2 | 72 | |||
FQ215132.1 | - | 6 - 7 | 998 | 72 | ||
FQ224976.1 | - | 6 - 7 | 1033 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
