Assay Details
Gene Symbol: Creb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006238050.1 | XP_006238112.1 | 6 - 7 | 852 | 136 | |
XM_008763672.1 | XP_008761894.1 | 7 - 8 | 656 | 136 | ||
GenBank mRNA | KJ817427.1 | - | 6 - 7 | 670 | 136 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
