Assay Details
Gene Symbol: Fbxl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107919.1 | NP_001101389.1 | 1 - 2 | 125 | 83 | |
XM_006237942.1 | XP_006238004.1 | 1 - 2 | 95 | 83 | ||
GenBank mRNA | BC161859.1 | - | 1 - 2 | 95 | 83 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC161859.1, NM_001107919.1, XM_006237942.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
