Assay Details
Gene Symbol: Fbxl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107919.1 | NP_001101389.1 | 6 - 7 | 1584 | 88 | |
XM_006237942.1 | XP_006238004.1 | 6 - 7 | 1591 | 88 | ||
GenBank mRNA | BC161859.1 | - | 6 - 7 | 1591 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
