Assay Details
Gene Symbol: Mmp16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080776.1 | NP_542954.1 | 2 - 3 | 714 | 125 | |
XM_006237921.2 | XP_006237983.1 | 2 - 3 | 780 | 125 | ||
GenBank mRNA | D63886.1 | - | 2 - 3 | 714 | 125 | |
D85509.1 | - | 2 - 3 | 714 | 125 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
