Assay Details
Gene Symbol: Rmdn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031663.1 | NP_001026833.1 | 7 - 8 | 730 | 68 | |
XM_006237933.2 | XP_006237995.1 | 6 - 7 | 773 | 68 | ||
GenBank mRNA | BC079473.1 | - | 7 - 8 | 736 | 68 | |
BC098713.1 | - | 7 - 8 | 730 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
