Assay Details
Gene Symbol: Cntfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003929.1 | NP_001003929.1 | 2 - 3 | 166 | 109 | |
XM_006238055.2 | XP_006238117.1 | 1 - 2 | 88 | 109 | ||
GenBank mRNA | S54212.1 | - | 2 - 3 | 166 | 109 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
